Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021602.hg.1 |
TC04001520.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021602.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021602.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021604.hg.1 |
TC04001520.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021604.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021604.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021600.hg.1 |
TC04001520.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021600.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021600.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180912.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180912.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180912.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180912.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180912.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180912.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180912.hg.1 |
TC0400011754.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400180916.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180916.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180916.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180916.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180916.hg.1 |
TC0400011754.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400180914.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180914.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180914.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180914.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180914.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180914.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180914.hg.1 |
TC0400011754.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000226730.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
ENST00000477645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011754.hg.1 |
NM_000586.3 |
Refseq |