Assay Details
Gene Symbol: KRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570378.2 | XP_005570435.1 | 3 - 4 | 415 | 111 | |
XM_005570379.2 | XP_005570436.1 | 3 - 4 | 483 | 111 | ||
XM_005570381.2 | XP_005570438.1 | 3 - 4 | 483 | 111 | ||
XM_015430533.1 | XP_015286019.1 | 3 - 4 | 483 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
