Assay Details
Gene Symbol: CISH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005547243.2 | XP_005547300.1 | 2 - 3 | 1245 | 64 | |
XM_005547244.2 | XP_005547301.1 | 2 - 3 | 414 | 64 | ||
XM_005547245.2 | XP_005547302.1 | 2 - 3 | 814 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
