Assay Details
Gene Symbol: GABARAPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285057.1 | NP_001271986.1 | 3 - 4 | 326 | 70 | |
GenBank mRNA | AB174202.1 | - | 3 - 4 | 326 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
