Assay Details
Gene Symbol: ELAPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283681.1 | NP_001270610.1 | 22 - 23 | 3082 | 93 | |
XM_015452080.1 | XP_015307566.1 | 22 - 23 | 3243 | 93 | ||
XM_015452084.1 | XP_015307570.1 | 21 - 22 | 3199 | 93 | ||
GenBank mRNA | AB169307.1 | - | 22 - 23 | 3082 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
