Assay Details
Gene Symbol: GABPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541937.2 | XP_005541994.1 | 1 - 2 | 589 | 100 | |
XM_005541938.2 | XP_005541995.1 | 1 - 2 | 589 | 100 | ||
XM_015457543.1 | XP_015313029.1 | 1 - 2 | 589 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
