Assay Details
Gene Symbol: C1QTNF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005558429.2 | XP_005558486.1 | 2 - 3 | 2424 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C1QTNF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005558429.2 | XP_005558486.1 | 2 - 3 | 2424 | 88 |
Set Membership: |
![]() ![]() ![]() ![]() |