Assay Details
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449302.1 | XP_015304788.1 | 38 - 39 | 8056 | 65 | |
XM_015449307.1 | XP_015304793.1 | 38 - 39 | 8056 | 65 | ||
XM_015449309.1 | XP_015304795.1 | 37 - 38 | 7914 | 65 | ||
XM_015449318.1 | XP_015304804.1 | 31 - 32 | 7111 | 65 | ||
XM_015449323.1 | XP_015304809.1 | 22 - 23 | 5348 | 65 |

More Information
Set Membership: |
![]() ![]() |
