Assay Details
Gene Symbol: FOXO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551534.2 | XP_005551591.1 | 3 - 4 | 2334 | 76 | |
GenBank mRNA | AB172276.1 | - | 1 - 2 | 1039 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
