Assay Details
Gene Symbol: CHRNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563230.1 | XP_005563287.1 | 3 - 4 | 703 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
