Assay Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284596.1 | NP_001271525.1 | 1 - 2 | 171 | 73 | |
GenBank mRNA | AB171181.1 | - | 1 - 2 | 171 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
