Assay Details
Gene Symbol: FAM131A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283598.1 | NP_001270527.1 | 4 - 5 | 536 | 64 | |
XM_005546565.2 | XP_005546622.1 | 5 - 6 | 495 | 64 | ||
XM_005546567.2 | XP_005546624.1 | 3 - 4 | 702 | 64 | ||
XM_015445325.1 | XP_015300811.1 | 4 - 5 | 756 | 64 | ||
GenBank mRNA | AB171769.1 | - | 4 - 5 | 536 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
