Assay Details
Gene Symbol: NTMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005580651.2 | XP_005580708.1 | 1 - 2 | 515 | 65 | |
XM_015436350.1 | XP_015291836.1 | 1 - 2 | 576 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
