Assay Details
Gene Symbol: POLR3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005562782.2 | XP_005562839.1 | 2 - 3 | 279 | 96 | |
XM_005562783.2 | XP_005562840.1 | 2 - 3 | 249 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
