Assay Details
Gene Symbol: SP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571005.2 | XP_005571062.1 | 2 - 3 | 274 | 77 | |
XM_005571006.2 | XP_005571063.1 | 2 - 3 | 176 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
