Assay Details
Gene Symbol: IRAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570388.2 | XP_005570445.1 | 20 - 21 | 2243 | 81 | |
XM_005570389.2 | XP_005570446.1 | 19 - 20 | 2210 | 81 | ||
XM_005570390.2 | XP_005570447.1 | 19 - 20 | 2162 | 81 | ||
XM_005570391.2 | XP_005570448.1 | 17 - 18 | 1952 | 81 | ||
XM_005570392.2 | XP_005570449.1 | 17 - 18 | 2067 | 81 | ||
XM_005570393.2 | XP_005570450.1 | 17 - 18 | 1997 | 81 | ||
XM_005570394.2 | XP_005570451.1 | 14 - 15 | 1038 | 81 | ||
XM_015430522.1 | XP_015286008.1 | 20 - 21 | 2240 | 81 | ||
XM_015430523.1 | XP_015286009.1 | 19 - 20 | 2207 | 81 | ||
XM_015430524.1 | XP_015286010.1 | 19 - 20 | 2180 | 81 | ||
XM_015430525.1 | XP_015286011.1 | 16 - 17 | 1919 | 81 | ||
XM_015430526.1 | XP_015286012.1 | 19 - 20 | 1631 | 81 | ||
XM_015430527.1 | XP_015286013.1 | 19 - 20 | 2181 | 81 | ||
XM_015430528.1 | XP_015286014.1 | 18 - 19 | 1754 | 81 | ||
XM_015430529.1 | XP_015286015.1 | 18 - 19 | 1598 | 81 | ||
XM_015430530.1 | XP_015286016.1 | 18 - 19 | 2148 | 81 | ||
XM_015430531.1 | XP_015286017.1 | 18 - 19 | 1940 | 81 | ||
XM_015430532.1 | XP_015286018.1 | 17 - 18 | 2000 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
