Assay Details
Gene Symbol: KRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570378.2 | XP_005570435.1 | 4 - 5 | 574 | 110 | |
XM_005570379.2 | XP_005570436.1 | 4 - 5 | 642 | 110 | ||
XM_015430533.1 | XP_015286019.1 | 4 - 5 | 642 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
