Assay Details
Gene Symbol: RAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567377.1 | XP_005567434.1 | 6 - 7 | 721 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
