Assay Details
Gene Symbol: NBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576512.2 | XP_005576569.1 | 7 - 8 | 607 | 71 | |
XM_015433977.1 | XP_015289463.1 | 7 - 8 | 607 | 71 | ||
XM_015433978.1 | XP_015289464.1 | 7 - 8 | 456 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015433978.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
