Assay Details
Gene Symbol: RPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579101.2 | XP_005579158.1 | 2 - 3 | 253 | 70 | |
GenBank mRNA | AB170681.1 | - | 2 - 3 | 199 | 70 | |
AB170967.1 | - | 2 - 3 | 188 | 70 | ||
AB172558.1 | - | 2 - 3 | 192 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
