Assay Details
Gene Symbol: HUNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549480.2 | XP_005549537.2 | 8 - 9 | 1509 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
