Assay Details
Gene Symbol: FGF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287283.1 | NP_001274212.1 | 3 - 4 | 986 | 99 | |
GenBank mRNA | AB063051.1 | - | 3 - 4 | 986 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
