Assay Details
Gene Symbol: ALKAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015434060.1 | XP_015289546.1 | 2 - 3 | 583 | 77 | |
XM_015434061.1 | XP_015289547.1 | 2 - 3 | 578 | 77 | ||
XM_015434062.1 | XP_015289548.1 | 2 - 3 | 582 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
