Assay Details
Gene Symbol: R3HCC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283748.1 | NP_001270677.1 | 8 - 9 | 2311 | 78 | |
GenBank mRNA | AB169220.1 | - | 8 - 9 | 2311 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
