Assay Details
Gene Symbol: HERC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555394.2 | XP_005555451.1 | 12 - 13 | 1720 | 99 | |
GenBank mRNA | AB168451.1 | - | 3 - 4 | 309 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
