Assay Details
Gene Symbol: KDELR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285233.1 | NP_001272162.1 | 2 - 3 | 296 | 104 | |
XM_005549079.2 | XP_005549136.1 | 2 - 3 | 357 | 104 | ||
GenBank mRNA | AB171648.1 | - | 2 - 3 | 296 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
