Assay Details
Gene Symbol: SEL1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015430092.1 | XP_015285578.1 | 3 - 4 | 442 | 70 | |
XM_015430093.1 | XP_015285579.1 | 3 - 4 | 318 | 70 | ||
XM_015430094.1 | XP_015285580.1 | 3 - 4 | 442 | 70 | ||
XM_015430095.1 | XP_015285581.1 | 3 - 4 | 442 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
