Assay Details
Gene Symbol: CHST9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005586983.2 | XP_005587040.1 | 6 | 2080 | 133 | |
XM_005586986.2 | XP_005587043.1 | 5 | 2042 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
