Assay Details
Gene Symbol: CIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005560193.2 | XP_005560250.1 | 5 - 6 | 1096 | 74 | |
XM_005560194.2 | XP_005560251.1 | 6 - 7 | 1113 | 74 | ||
XM_015453157.1 | XP_015308643.1 | 4 - 5 | 551 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
