Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005539643.2 | XP_005539700.1 | 14 - 15 | 1906 | 76 | |
XM_005539646.2 | XP_005539703.1 | 14 - 15 | 1906 | 76 | ||
XM_015433592.1 | XP_015289078.1 | 14 - 15 | 2003 | 76 | ||
XM_015433624.1 | XP_015289110.1 | 14 - 15 | 2003 | 76 | ||
XM_015433665.1 | XP_015289151.1 | 13 - 14 | 1877 | 76 | ||
XM_015433704.1 | XP_015289190.1 | 14 - 15 | 2003 | 76 | ||
XM_015433732.1 | XP_015289218.1 | 14 - 15 | 2003 | 76 | ||
XM_015433787.1 | XP_015289273.1 | 14 - 15 | 2003 | 76 | ||
XM_015433878.1 | XP_015289364.1 | 14 - 15 | 2003 | 76 | ||
XM_015433922.1 | XP_015289408.1 | 14 - 15 | 2003 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
