Assay Details
Gene Symbol: ACTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575577.2 | XP_005575634.1 | 6 - 7 | 769 | 80 | |
GenBank mRNA | AB168686.1 | - | 8 - 9 | 1319 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
