Assay Details
Gene Symbol: RLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543842.2 | XP_005543899.1 | 7 - 8 | 1120 | 81 | |
XM_005543843.2 | XP_005543900.1 | 4 - 5 | 565 | 81 | ||
XM_015437825.1 | XP_015293311.1 | 4 - 5 | 596 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
