Assay Details
Gene Symbol: BRCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005595046.2 | XP_005595103.1 | 7 - 8 | 652 | 86 | |
XM_005595047.2 | XP_005595104.1 | 7 - 8 | 653 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
