Assay Details
Gene Symbol: UBE2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283170.1 | NP_001270099.1 | 1 - 2 | 73 | 64 | |
XM_005545650.2 | XP_005545707.1 | 1 - 2 | 1271 | 64 | ||
GenBank mRNA | AB178995.1 | - | 1 - 2 | 73 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
