Assay Details
Gene Symbol: CCL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287694.1 | NP_001274623.1 | 2 - 3 | 178 | 74 | |
GenBank mRNA | EU499667.1 | - | 2 - 3 | 178 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
