Assay Details
Gene Symbol: CXCL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005565094.2 | XP_005565151.1 | 2 - 3 | 280 | 59 | |
XM_005565095.2 | XP_005565152.1 | 2 - 3 | 280 | 59 | ||
XM_005565096.2 | XP_005565153.1 | 2 - 3 | 279 | 59 | ||
XM_005565097.2 | XP_005565154.1 | 2 - 3 | 279 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
