Assay Details
Gene Symbol: HRH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569520.2 | XP_005569577.1 | 2 - 3 | 486 | 71 | |
XM_005569521.2 | XP_005569578.1 | 2 - 3 | 433 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
