Assay Details
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284699.1 | NP_001271628.1 | 3 - 4 | 359 | 146 | |
GenBank mRNA | D63352.1 | - | 3 - 4 | 359 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
