Assay Details
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285277.1 | NP_001272206.1 | 1 - 2 | 217 | 109 | |
GenBank mRNA | AB000513.1 | - | 1 - 2 | 217 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
