Assay Details
Gene Symbol: PEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563584.2 | XP_005563641.1 | 2 - 3 | 302 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005563584.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
