Assay Details
Gene Symbol: NQO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592390.2 | XP_005592447.1 | 3 - 4 | 807 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
