Assay Details
Gene Symbol: VIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284705.1 | NP_001271634.1 | 4 - 5 | 1031 | 76 | |
GenBank mRNA | AB169770.1 | - | 4 - 5 | 1072 | 76 | |
AB170255.1 | - | 4 - 5 | 1031 | 76 |
