Assay Details
Gene Symbol: FAM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005594988.2 | XP_005595045.1 | 1 - 2 | 205 | 76 | |
XM_005594989.2 | XP_005595046.1 | 1 - 2 | 204 | 76 | ||
XM_015444267.1 | XP_015299753.1 | 1 - 2 | 205 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
