Assay Details
Gene Symbol: PDCD2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588785.2 | XP_005588842.1 | 3 - 4 | 370 | 72 | |
XM_005588786.2 | XP_005588843.1 | 3 - 4 | 370 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
