Assay Details
Gene Symbol: CDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544549.2 | XP_005544606.1 | 3 - 4 | 505 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
