Assay Details
Gene Symbol: GJB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005585446.2 | XP_005585503.1 | 2 - 3 | 1201 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005585446.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
