Assay Details
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563546.2 | XP_005563603.1 | 3 - 4 | 228 | 79 | |
XM_005563548.2 | XP_005563605.2 | 3 - 4 | 295 | 79 | ||
XM_015454971.1 | XP_015310457.1 | 3 - 4 | 228 | 79 | ||
XM_015454972.1 | XP_015310458.1 | 2 - 3 | 195 | 79 | ||
XM_015454974.1 | XP_015310460.1 | 2 - 3 | 196 | 79 | ||
XM_015454975.1 | XP_015310461.1 | 2 - 3 | 170 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015454975.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
