Assay Details
Gene Symbol: ARL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005572006.2 | XP_005572063.1 | 4 - 5 | 1093 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
