Assay Details
Gene Symbol: HOPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555220.2 | XP_005555277.1 | 1 - 2 | 38 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005555220.2 transcripts.
Set Membership: |
![]() ![]() |
