Assay Details
Gene Symbol: TEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005562418.2 | XP_005562475.1 | 2 - 3 | 436 | 115 | |
XM_005562420.2 | XP_005562477.1 | 2 - 3 | 446 | 115 | ||
XM_005562421.2 | XP_005562478.1 | 2 - 3 | 427 | 115 | ||
XM_015454282.1 | XP_015309768.1 | 2 - 3 | 441 | 115 | ||
XM_015454283.1 | XP_015309769.1 | 2 - 3 | 432 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
